Learn More
VITLAB™ Piston-Operated Micropipettes
Product Code. 11938121
Click to view available options
Volume (Metric):
100 to 1000 μL
1000 to 10,000 μL
500 to 5000 μL
Accuracy:
±30μL
±6μL
±60μL
Unit Size:
Each
Product Code. 11938121
Brand: VITLAB™ 1641008
Only null left
Add to basket
Add to basket
The VITALAB™ piston-operated pipettes are the ideal manual pipettes for demanding laboratory applications with a robust design, ergonomic shape and simple operation.
  • Completely autoclavable at 121°C acc. to DIN EN 285
  • Highly accurate with simple calibration for long-lasting reliability
  • Large, central pipetting button provides a uniform and smooth movement of the piston
  • Ergonomic eject button for rapid tip replacement
  • 4-digit volume display with integrated zoom function and vertical arrangement of the numbers ensures optimal readability at all times
  • Desired volume can be set by rotating the volume setting wheel with ease and precision
  • Integrated calibration function allows adjustment without tools directly in the laboratory
  • Corrosion-resistant piston and ejector ensure long product life
  • Includes micropipette, silicone grease, sample bag with pipette tips, quality certificate and operating manual
Specifications
Accuracy ±6μL
Autoclavable Autoclavable
Type Piston Operated Micropipette
Volume (Metric) 100 to 1000 μL
Display 4-digit
Includes Micropipette, silicone grease, sample bag with pipette tips, quality certificate, operating manual