Learn More
Thermo Scientific™ Micrococcal nuclease, Staphylococcus aureus
Single and double-stranded DNA and RNA endonuclease which yields 3' nucleotides.
Brand: Thermo Scientific™ J70196.Z^
Additional Details : CAS : 9013-53-0 Weight : 1.00000kg
Description
- Specific for single-stranded nucleic acids and shows preference for AU or AT-rich regions
- Complete digestion yields mono- and dinucleotides
- MDL: MFCD00131731
Specifications
9013-53-0 | |
E.C.3.1.31.1 | |
15 kU |
Keep cold | |
Staphylococcus aureus | |
Solid |