missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICB Monoclonal Antibody (04), Invitrogen™

Mouse Monoclonal Antibody
258.00 € - 449.00 €
Specifications
| Antigen | MICB |
|---|---|
| Clone | 04 |
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17719753
|
Invitrogen
MA543393 |
50 μL |
258.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17749553
![]() |
Invitrogen
MA542999 |
100 μL |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specificity: Human MICB
MICB encodes the highly polymorphic MHC (HLA) class I chain-related gene B. The protein product is expressed on the cell surface. It is thought that MICB functions as a stress-induced antigen that is broadly recognized by intestinal epithelial gamma delta T cells. MICB is broadly recognized by NK cells and T cells with NKG2D receptor on their surface. The complex NKG2D-MICB results in MICB expressing cytolytic T cells and NK cells against epithelial tumor cells.Specifications
| MICB | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| MHC class I chain-related protein B; MHC class I mic-B antigen; MHC class I polypeptide-related sequence B; MHC class I-like located near the LRC, 2; MHC class I-like molecule PERB11.2-IMX; MHC I like leukocyte 2; MICB; MIC-B; Mill2; Mill2 gene for MHC class I-like located near the LRC, 2, exon 3, partial cds, strain:LEW.1lm1; PERB11.2; sMICB; soluble MICB; stress inducible class I homolog | |
| MICB | |
| IgG1 | |
| Protein A | |
| Antibody |
| 04 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q29980 | |
| 4277 | |
| Recombinant Human MICB Protein. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| MICB |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title