missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MIC-1 Polyclonal Antibody

Description
Antibody detects endogenous levels of total MIC1.
Colon cancer-associated protein Mic1.
Specifications
Specifications
| Antigen | MIC-1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Rmc1 |
| Gene Accession No. | Q8VC42, Q96DM3 |
| Gene Alias | 2400010D15Rik; 3110002H16Rik; C18orf8; chromosome 18 open reading frame 8; colon cancer-associated protein Mic1; colon cancer-associated protein Mic1 homolog; HsT2591; macrophage inhibitory cytokine 1; Mic1; Mic-1; Regulator of MON1-CCZ1 complex; RGD1311805; RIKEN cDNA 3110002H16 gene; RMC1; similar to RIKEN cDNA 2400010D15; uncharacterized protein C18orf8; uncharacterized protein C18orf8 homolog; WD repeat-containing protein 98; WDR98 |
| Gene Symbols | Rmc1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?