missing translation for 'onlineSavingsMsg'
Learn More

MF63, MedChemExpress

Product Code. 16426156
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16426156

Brand: Medchem Express HY13283

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MF63 is a selective mPGES-1 inhibitor with an IC50 of 0.9 nM and 1.3 nM for pig mPGES-1 and human mPGES-1 enzyme, respectively.

MF63 is a selective mPGES-1 inhibitor with an IC50 of 0.9 nM and 1.3 nM for pig mPGES-1 and human mPGES-1 enzyme, respectively.

Specifications

Chemical Name or Material MF63
CAS 892549-43-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C23H11ClN4
Quantity 100 mg
Solubility Information DMSO : ≥ 43 mg/mL (113.51 mM)
SMILES ClC1=CC(C2=C3C=CC=C2)=C(C4=C3N=C(C5=C(C=CC=C5C#N)C#N)N4)C=C1
Molecular Weight (g/mol) 378.81
Formula Weight 378.81
Percent Purity 99.05%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.