Learn More
Mettler Toledo™ Excellence RM Digital Benchtop Refractometers
Product Code. 10468845 Shop All Mettler Toledo Products
Change view
Click to view available options
RM40; 4-place; 5.0° to 100.0°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10468845 RM40; 4-place; 5.0° to 100.0°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10468845 Supplier Mettler Toledo™ Supplier No. 51337003
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Benchtop refractometers allow you to determine the refractive index, BRIX, HFCS, or user-defined concentrations of a wide variety of liquids quickly and accurately. Mettler Toledo Excellence RM Digital Benchtop Refractometer has an in-built Peltier temperature control which provides both reliable temperature stability and rapid heating/cooling rates.
  • Inert sapphire prism facilitates measurement of thin, highly volatile, dark, or viscous liquids
  • Removable top section provides ease of cleaning with simple tap water without the need for tools
  • Plug-and-play connection of accessories such as the printer, barcode scanner, LogStraight™ biometric fingerprint reader, and ErgoSens™ infrared sensor for hands-free operation
  • Assisted sample loading and clearing are possible through the addition of FillPal™ and DryPal™ units
  • Optional EQPac (Equipment Qualification Package) provides comprehensive, detailed qualification guidance (IQ/OQ/PQ) to those working in regulated environments
Expand Your Scope of Measurement
  • LiquiPhysics™ allows you to add parameters such as pH, color, and density/specific gravity to your refractive index readings
  • Simply connect a compatible SevenCompact™ pH or conductivity meter, colorimeter, or density cell to the RM module and define the method
  • Optional LabX™ LiquiPhysics™ Express software captures and manages all of these data points under a single platform
  • LabX™ also fully supports your CFR 21.11 requirements, provides user management, audit trail, flexible data export options, and fully customized reports
Reliable Standards
  • Mettler Toledo Certified Combined Density and Refractive Index Standards reduce risk, uncertainty, and error-prone sample transfer Standards come complete with certificates including an easy-to-scan barcode containing all information necessary for calibration
  • Barcodes save you valuable time, reduce data entry error, and improve traceability
TRUSTED_SUSTAINABILITY
Specifications
Scale Range 0 to 100 Brix
Refractive Index 1.32 to 1.7
Description Model RM40
Accuracy 0.0001, 0.1% Brix
Measurement Time 20 sec.
Temperature Range (English) 41°F to 212°F
Temperature Range (Metric) 5°C to 100°C
Frequency 50/60 Hz