missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human METTL7B Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0758
Additional Details : Weight : 0.01000kg
Description
MettL7B Synthetic Peptide, PEP-0758, from Invitrogen.
This peptide corresponds to 13 amino acids near the amino terminus of human MettL7B.PEP-0758 can be used as a blocking peptide with polyclonal antibody PA5-20638.Specifications
196410 | |
purified | |
Human METTL7B | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q6UX53 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
METTL7B | |
0.02% sodium azide | |
50 μg | |
PBS |