Learn More
METTL7A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Product Code. 16856934 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16856934 20 μL 20µL
16846934 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16856934 Supplier Proteintech Supplier No. 170921AP20UL
Please to purchase this item. Need a web account? Register with us today!
Rabbit Polyclonal Antibody
The METTL7A antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human METTL7A. This antibody recognizes human, mouse, rat antigen. The METTL7A antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Antigen METTL7A
Applications Western Blot, Immunohistochemistry, ELISA
Classification Polyclonal
Concentration 267 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 25840
Target Species Human, Rat, Mouse
Isotype IgG