missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ METTL10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569581
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Protein-lysine methyltransferase that selectively catalyzes the trimethylation of EEF1A at 'Lys-318'. [UniProt].
Specifications
| METTL10 | |
| Polyclonal | |
| Unconjugated | |
| EEF1AKMT2 | |
| 2010208K18Rik; C10orf138; EEF1A lysine methyltransferase 2; EEF1AKMT2; Em:AC068896.3; methyltransferase like 10; methyltransferase-like protein 10; Mettl10; protein-lysine N-methyltransferase Mettl10; RGD1306300 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 72096 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9D853 | |
| EEF1AKMT2 | |
| synthetic peptide directed towards the middle region of mouse METTL10 (aa 151-200). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction