Learn More
Metrohm 876 Manual Titrator Plus Liquid Handling Device
Description
Includes
Stirring bars (16mm/25mm), support stand, clamping ring, 400mm support rod, electrode holder, manual dosing controller, USB MINI - USB A adapter, hex keys (2mm/5mm), wrench and exchange unit (20mL)
- Manual titrator with result calculation for manual titrations and dosings
- With intelligent exchange unit (20mL), manual dosing controller and 801 magnetic stirrer including stand
- Dialogue languages: DE, EN, ES, FR
- Increments per cylinder volume: 10'000
- With printer connection (USB), remote interface, RS232 connection for balance, remote control via RS232 (method load-up, start, stop) and pulse mode
Dispensing Modes
Manual Dosing (DOS): Push-button dosing; Particularly suitable for carrying out manual titrations with a color indicator; From the added volume the results can be calculated automatically and the report printed out; Different calculation parameters can be defined beforehand
Extended Dosing (XDOS): Adding a fixed volume: enter volume and dosing rate; Time-controlled dosing: enter volume and time; Constant dosing rate: enter dosing rate and time
Manual titration
Tandem dosing
Specifications
Specifications
| For Use With (Equipment) | Titrino/Titrino+ titrators |
| Includes | Stirring bars (16mm/25mm), support stand, clamping ring, 400mm support rod, electrode holder, manual dosing controller, USB MINI - USB A adapter, hex keys (2mm/5mm), wrench and exchange unit (20mL) |
| Model | 876 Manual Titrator Plus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.