missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metoprolol succinate, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
Description
Metoprolol succinate is an orally active, selective β1-adrenoceptor antagonist. Metoprolol succinate shows anti-inflammation, antitumor and anti-angiogenic properties.
Specifications
Specifications
| Chemical Name or Material | Metoprolol succinate |
| CAS | 98418-47-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C34H56N2O10 |
| Quantity | 50 mg |
| Solubility Information | DMSO : 16.67 mg/mL (25.54 mM; Need ultrasonic) |
| SMILES | OC(CNC(C)C)COC1=CC=C(CCOC)C=C1.OC(CNC(C)C)COC2=CC=C(CCOC)C=C2.OC(CCC(O)=O)=O |
| Molecular Weight (g/mol) | 652.82 |
| Formula Weight | 652.82 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction