Learn More
Methylurea-d6, CDN
Specifications
Specifications
| Chemical Name or Material | N-Methylurea-d6 |
| CAS | 56683-43-3 |
| Molecular Formula | C2 D6 N2 O |
| Quantity | 0.25 g |
| Synonym | Urea-d3, methyl-d3- (9 CI), N-Methylurea-d6, Methylurea-D6, Methyl-d3-urea-d3, 1-(Methyl-d3)urea-d3 |
| InChI Formula | InChI=1 S/C2H6N2O/c1-4-2(3)5/h1H3,(H3,3,4,5)/i1D3/hD3 |
| SMILES | [2 H]N([2 H])C(=O)N([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,3-trideuterio-3-(trideuteriomethyl)urea |
| Molecular Weight (g/mol) | 80.1188 |
| Formula Weight | 80.0857 g/mol |
| Show More |