missing translation for 'onlineSavingsMsg'
Learn More

Methyltriphenylphosphonium chloride, 97%, Thermo Scientific Chemicals

Product Code. 15423398
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15423398 25 g 25g
15403398 1 g 1g
15413398 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15423398 Supplier Thermo Scientific Alfa Aesar Supplier No. H56851.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Methyltriphenylphosphonium chloride is used as a phase transfer catalyst in synthetic chemistry. It acts as a catalyst for regio-controlled ring-opening polymerization of substituted epoxides. It is utilized for controlling the corrosion iron. It acts as a reactant for intramolecular nitrone cycloaddition. In addition to this, it is used for neoplasm inhibition through antimitochondrial effect.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyltriphenylphosphonium chloride is used as a phase transfer catalyst in synthetic chemistry. It acts as a catalyst for regio-controlled ring-opening polymerization of substituted epoxides. It is utilized for controlling the corrosion iron. It acts as a reactant for intramolecular nitrone cycloaddition. In addition to this, it is used for neoplasm inhibition through antimitochondrial effect.

Solubility
Soluble in methanol.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1031-15-8
Molecular Formula C19H18ClP
Molecular Weight (g/mol) 312.78
MDL Number MFCD00797851
InChI Key QRPRIOOKPZSVFN-UHFFFAOYSA-M
Synonym methyltriphenylphosphonium chloride, methyl triphenyl phosphonium chloride, phosphonium, methyltriphenyl-, chloride, methyltriphenylphosphanium chloride, methyltriphenylphosphoniumchloride, methyl triphenylphosphoniumchloride, methyl triphenylphosphonium chloride, methyl triphenyl phosphanium chloride
PubChem CID 9879809
IUPAC Name methyl(triphenyl)phosphanium;chloride
SMILES [Cl-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 221°C (decomposition)
Linear Formula CH3P(C6H5)3Cl
Quantity 25 g
UN Number UN3077
Sensitivity Hygroscopic
Solubility Information Soluble in methanol.
Formula Weight 312.78
Percent Purity 97%
Chemical Name or Material Methyltriphenylphosphonium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.