missing translation for 'onlineSavingsMsg'
Learn More

Methyleo-toluate, 99%, ACROS Organics™

Product Code. 10413361 Shop All Thermo Scientific Chemicals Products
Click to view available options
:
25g; Glass bottle
This item is not returnable. View return policy

Product Code. 10413361

Brand: Thermo Scientific Acros 414950250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 89-71-4
Color Colorless
Density 1.0700g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C9H10O2
Linear Formula CH3C6H4COOCH3
Refractive Index 1.5180 to 1.5200 (20°C, 589nm)
MDL Number MFCD00008428
Quantity 25 g
Synonym methyl o-toluate, methyl orthotoluate, o-toluic acid, methyl ester, methyl o-methylbenzoate, methyl toluate, benzoic acid, 2-methyl-, methyl ester, 2-methylbenzoic acid methyl ester, o-toluic acid methyl ester, methyl-o-methyl benzoate, 2-methyl methylbenzoate
InChI Key WVWZECQNFWFVFW-UHFFFAOYSA-N
SMILES CC1=CC=CC=C1C(=O)OC
IUPAC Name methyl 2-methylbenzoate
Molecular Weight (g/mol) 150.177
PubChem CID 33094
Formula Weight 150.18
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Methyl o-toluate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.