Learn More
Methylamine, extra pure, 40 wt% solution in water
Product Code. p-7214324 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
10 L
Unit Size:
1 L
10 L
2.5 L
Product Code. 10687282
Brand: Thermo Scientific Acros 126230010
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 74-89-5 | CH5N | 31.06 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 74-89-5
Molecular Formula CH5N
Molecular Weight (g/mol) 31.06
InChI Key BAVYZALUXZFZLV-UHFFFAOYSA-N
IUPAC Name methanamine
SMILES CN
Specifications
Melting Point -38.0°C
Density 0.9000 g/mL
Boiling Point 48.0°C
Flash Point -15°C
Packaging Glass bottle
Linear Formula CH3NH2
Refractive Index 1.3650 to 1.3700
Quantity 1 L
Beilstein 04, 32
Merck Index 15, 6088
Specific Gravity 0.9
Solubility Information Solubility aq. soln.: freely soluble.
Formula Weight 31.06
Grade Extra Pure
Chemical Name or Material Methylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methylamine, 40 wt% solution in water
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.