missing translation for 'onlineSavingsMsg'
Learn More

Methyl valerate, 99%

Product Code. 11476596
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11476596

Brand: Thermo Scientific Alfa Aesar B24828.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 624-24-8 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-24-8
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009478
InChI Key HNBDRPTVWVGKBR-UHFFFAOYSA-N
Synonym methyl valerate, pentanoic acid, methyl ester, methyl n-valerate, methyl valerianate, valeric acid, methyl ester, valeric acid methyl ester, unii-zw21jjj9vn, pentanoic acid methyl ester, fema no. 2752, zw21jjj9vn
PubChem CID 12206
IUPAC Name methyl pentanoate
SMILES CCCCC(=O)OC

Specifications

Melting Point -91°C
Density 0.885
Boiling Point 126°C to 128°C
Flash Point 22°C (71°F)
Refractive Index 1.397
Quantity 500 g
UN Number UN3272
Beilstein 1741905
Formula Weight 116.16
Percent Purity 99%
Chemical Name or Material Methyl valerate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.