Learn More
Methyl thiazolidine-2-carboxylate hydrochloride, 98%
Product Code. 11382966
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11382966
Brand: Thermo Scientific Alfa Aesar L10740.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 50703-06-5 | C5H10ClNO2S | 183.65 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 50703-06-5
Molecular Formula C5H10ClNO2S
Molecular Weight (g/mol) 183.65
MDL Number MFCD00068022
InChI Key YEFOSJYXVLNYSL-UHFFFAOYNA-N
Synonym 2-thiazolidinecarboxylic acid methyl ester, 2-thiazolidinecarboxylic acid, methyl ester, thiazolidine-2-carboxylic acid methyl ester, methyl thiazolidine-2-carboxylate, +-thiazolidine-2-carboxylic acid methyl ester, +/--thiazolidine-2-carboxylic acid methyl ester, 2-thiazolidinecarboxylic acid, methyl ester 6ci,8ci,9ci
PubChem CID 2737507
IUPAC Name methyl 1,3-thiazolidine-2-carboxylate
SMILES [H+].[Cl-].COC(=O)C1CSCN1
Specifications
Melting Point ∽158°C (decomposition)
Quantity 1 g
Beilstein 3628640
Formula Weight 183.66
Percent Purity 98%
Chemical Name or Material Methyl thiazolidine-2-carboxylate hydrochloride