Learn More
Methyl (S)-Pyroglutamate, TRC
Product Code. 30465033
Change view
Click to view available options
Quantity:
1 g
2 g
5 g
Unit Size:
1g
2g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30465033 5 g 5g
30466330 2 g 2g
30470916 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30465033 Supplier Toronto Research Chemicals Supplier No. TRCM3259485G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4931-66-2
Molecular Formula C6 H9 N O3
Molecular Weight (g/mol) 143.14
IUPAC Name methyl (2S)-5-oxopyrrolidine-2-carboxylate
SMILES COC(=O)[C@@H]1CCC(=O)N1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C6H9NO3/c1-10-6(9)4-2-3-5(8)7-4/h4H,2-3H2,1H3,(H,7,8)/t4-/m0/s1
Formula Weight 143.0582
Chemical Name or Material Methyl (S)-Pyroglutamate