Learn More
Methyl o-toluate, 99%
Product Code. p-7022782
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. 11467647
Brand: Thermo Scientific Alfa Aesar A15695.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 89-71-4 | C9H10O2 | 150.177 g/mol
It has been used in the preparation of 2-(azidomethyl)benzoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It has been used in the preparation of 2-(azidomethyl)benzoic acid.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 89-71-4
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00008428
InChI Key WVWZECQNFWFVFW-UHFFFAOYSA-N
Synonym methyl o-toluate, methyl orthotoluate, o-toluic acid, methyl ester, methyl o-methylbenzoate, methyl toluate, benzoic acid, 2-methyl-, methyl ester, 2-methylbenzoic acid methyl ester, o-toluic acid methyl ester, methyl-o-methyl benzoate, 2-methyl methylbenzoate
PubChem CID 33094
IUPAC Name methyl 2-methylbenzoate
SMILES CC1=CC=CC=C1C(=O)OC
Specifications
Density 1.072
Boiling Point 207°C to 208°C
Flash Point 82°C (179°F)
Refractive Index 1.519
Quantity 25 g
Beilstein 2041846
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material Methyl o-toluate
compliance-icons
Product Identifier
  • Methyl o-toluate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Methyl o-toluate