Learn More
Methyl N-Hydroxy-2-methylphenylcarbamate, TRC
Product Code. 30466475
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30466475 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30466475 Supplier Toronto Research Chemicals Supplier No. TRCM2587151G
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl N-Hydroxy-2-methylphenylcarbamate
CAS 151830-35-2
Molecular Formula C9H11NO3
Quantity 1 g
InChI Formula InChI=1S/C9H11NO3/c1-7-5-3-4-6-8(7)10(12)9(11)13-2/h3-6,12H,1-2H3
SMILES COC(=O)N(O)c1ccccc1C
Molecular Weight (g/mol) 181.19
Formula Weight 181.0739
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.