Learn More
Methyl Mycophenolate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl (4E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroiso-benzofuran-5-yl)-4-methylhex-4-enoate |
| CAS | 31858-66-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18 H22 O6 |
| Quantity | 100 mg |
| Synonym | Methyl (4E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoate, 4-Hexenoic acid','6-(1,3-dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-','methyl ester','(4E)-, 4-Hexenoic acid','6-(1,3-dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-','methyl ester','(E)-, 4-Hexenoic acid','6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl)-4-methyl-','methyl ester','(E)- (8CI), Methyl mycophenolate, Mycophenolic acid methyl ester, Methyl (4E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroiso-benzofuran-5-yl)-4-methylhex-4-enoate, Mycophenolate Mofetil Imp. E (EP), Methyl Mycophenolate (EP Impurity E) |
| InChI Formula | InChI=1S/C18H22O6/c1-10(6-8-14(19)22-3)5-7-12-16(20)15-13(9-24-18(15)21)11(2)17(12)23-4/h5,20H,6-9H2,1-4H3/b10-5+ |
| SMILES | COC(=O)CC\C(=C\Cc1c(O)c2C(=O)OCc2c(C)c1OC)\C |
| IUPAC Name | methyl (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoate |
| Molecular Weight (g/mol) | 334.36 |
| Show More |