missing translation for 'onlineSavingsMsg'
Learn More

Methyl Methanethiosulfonate, TRC

Product Code. 17901845
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17901845 100 mg 100mg
30135879 5 g 5g
30137117 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17901845 Supplier Toronto Research Chemicals Supplier No. TRCM321500100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2949-92-0
InChI Formula InChI=1S/C2H6O2S2/c1-5-6(2,3)4/h1-2H3
IUPAC Name methylsulfonylsulfanylmethane
Molecular Formula C2 H6 O2 S2
Molecular Weight (g/mol) 126.2
Chemical Name or Material Methyl Methanethiosulfonate

Specifications

Quantity 100 mg
Synonym S-Methyl Methanethiosulfonate
SMILES CSS(=O)(=O)C
Formula Weight 125.9809
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.