missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl-Lysine Antibody [Alexa Fluor« 750], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35817AF750
This item is not returnable.
View return policy
Description
Methyl-Lysine Polyclonal antibody specifically detects Methyl-Lysine in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Methyl-Lysine | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| Alexa Fluor 750 | |
| Dimethylysine, Methyl lysine, methyllysine, monomethyl lysine, N epsilon dimethyl lysine, N epsilon mono methyllysine, N epsilon trimethyl lysine, N(epsilon) trimethyl lysine, Trimethyl lysine | |
| A synthetic peptide corresponding to a sequence containing monomethylated K. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction