Learn More
Methyl Desoxycholate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl Desoxycholate |
| CAS | 3245-38-3 |
| Molecular Formula | C25 H42 O4 |
| Quantity | 250 mg |
| Synonym | Cholic Acid Impurity 95 |
| InChI Formula | InChI=1S/C25H42O4/c1-15(5-10-23(28)29-4)19-8-9-20-18-7-6-16-13-17(26)11-12-24(16,2)21(18)14-22(27)25(19,20)3/h15-22,26-27H,5-14H2,1-4H3/t15-,16-,17-,18+,19-,20+,21+,22+,24+,25-/m1/s1 |
| SMILES | COC(=O)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C |
| IUPAC Name | methyl (4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate |
| Molecular Weight (g/mol) | 406.6 |
| Formula Weight | 406.3083 |