Learn More
Methyl-d3 Formate, CDN
Product Code. 30317260
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30317260 5 g 5g
30326595 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30317260 Supplier CDN Supplier No. CDND02485G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl Formate-D3 (methyl-D3)
CAS 23731-39-7
Molecular Formula C2 D3 H O2
Quantity 5 g
Synonym Methan-d3-ol, formate (9 CI), Formic acid, methyl-d3 ester (6 CI, 7 CI, 8 CI), Methyl-d3 formate, Formic acid methyl-d3 ester, Methyl formate-D3 (methyl-D3)
InChI Formula InChI=1 S/C2H4O2/c1-4-2-3/h2H,1H3/i1D3
SMILES [2 H]C([2 H])([2 H])OC=O
IUPAC Name trideuteriomethyl formate
Molecular Weight (g/mol) 63.0705
Formula Weight 63.04 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity