missing translation for 'onlineSavingsMsg'
Learn More

Methyl α-D-mannopyranoside, ≥99%, Fisher BioReagents™

Product Code. 10286094
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Amber Glass
Unit Size:
100g
10g
25g
500g
This item is not returnable. View return policy

Product Code. 10286094

Brand: Fisher BioReagents BP253025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Methyl α-D-mannopyranoside, ≥99%

  • Methyl α-D-mannopyranoside is a lectin inhibitory sugar
  • Competitive inhibitor of the binding of mannose by E. coli
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-04-9
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00063262
InChI Key HOVAGTYPODGVJG-UHFFFAOYNA-N
Synonym methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
PubChem CID 101798
ChEBI CHEBI:43943
SMILES COC1OC(CO)C(O)C(O)C1O

Specifications

Melting Point 193°C to 196°C
Color White
Loss on Drying < 0.5%
Assay Percent Range 99%
Packaging Amber Glass
Quantity 25 g
Specific Rotation 76 to 85° (+ or -)
IUPAC Name 2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Formula Weight 194.2
Percent Purity ∼99%
Physical Form Powder or Solid
Chemical Name or Material Methyl α-D-Mannopyranoside
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.