Learn More
Methyl blue, MP Biomedicals™
Product Code. 11407992 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11407992 5 g 5g
11452546 10 g 10g
11417992 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11407992 Supplier MP Biomedicals Supplier No. 0215264305
Please to purchase this item. Need a web account? Register with us today!
Chemical Identifiers
CAS 28631-66-5
Molecular Formula C32H27N3
Molecular Weight (g/mol) 453.59
MDL Number MFCD00036130
InChI Key LBLFVSANTXZPEX-UHFFFAOYSA-N
Synonym Acid blue 93, Aniline blue
PubChem CID 131675892
IUPAC Name 2-methyl-4-{[4-(phenylamino)phenyl][4-(phenylimino)cyclohexa-2,5-dien-1-ylidene]methyl}aniline
SMILES CC1=CC(=CC=C1N)C(C1=CC=C(NC2=CC=CC=C2)C=C1)=C1C=CC(C=C1)=NC1=CC=CC=C1
Specifications
Formula Weight 799.8
Color Black/Purple to Blue
Quantity 5 g
Physical Form Powder
Chemical Name or Material Methyl blue