missing translation for 'onlineSavingsMsg'
Learn More

Methyl Benzenesulfonate, TRC

Product Code. 30010743
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010743 100 mg 100mg
30006948 1 g 1g
30003487 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010743 Supplier Toronto Research Chemicals Supplier No. TRCM288890100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-18-2
InChI Formula InChI=1S/C7H8O3S/c1-10-11(8,9)7-5-3-2-4-6-7/h2-6H,1H3
IUPAC Name methyl benzenesulfonate
Molecular Formula C7H8O3S
Molecular Weight (g/mol) 172.2
Chemical Name or Material Methyl Benzenesulfonate

Specifications

Quantity 100 mg
Synonym Methyl Benzenesulphonate, Benzenesulfonic acid, methyl ester (6CI,7CI,8CI,9CI), 20ND3-5, Methyl benzenesulfonate, NSC 3214
SMILES COS(=O)(=O)c1ccccc1
Formula Weight 172.0194
Percent Purity >95%
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.