Learn More
Methyl 6-Aminopyridine-3-carboxylate, TRC
Product Code. 30481948
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481948 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30481948 Supplier Toronto Research Chemicals Supplier No. TRCM2868551G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl 6-Aminopyridine-3-carboxylate
CAS 36052-24-1
Molecular Formula C7 H8 N2 O2
Quantity 1 g
Synonym 3-Pyridinecarboxylic acid','6-amino-','methyl ester, Nicotinic acid','6-amino-','methyl ester (6CI,8CI), 2-Amino-5-(methoxycarbonyl)pyridine, 5-Methoxycarbonyl-2-Aminopyridine, 6-Aminonicotinic acid methyl ester, 6-Aminopyridine-3-carboxylic acid methyl ester, Methyl 2-aminopyridine-5-carboxylate, Methyl 6-aminonicotinate, Methyl 6-aminopyridine-3-carboxylate
InChI Formula InChI=1S/C7H8N2O2/c1-11-7(10)5-2-3-6(8)9-4-5/h2-4H,1H3,(H2,8,9)
SMILES COC(=O)c1ccc(N)nc1
IUPAC Name methyl 6-aminopyridine-3-carboxylate
Molecular Weight (g/mol) 152.15
Formula Weight 152.0586