Learn More
Methyl 6-Aminopyridine-3-carboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 6-Aminopyridine-3-carboxylate |
| CAS | 36052-24-1 |
| Molecular Formula | C7 H8 N2 O2 |
| Quantity | 2.5 g |
| Synonym | 3-Pyridinecarboxylic acid','6-amino-','methyl ester, Nicotinic acid','6-amino-','methyl ester (6CI,8CI), 2-Amino-5-(methoxycarbonyl)pyridine, 5-Methoxycarbonyl-2-Aminopyridine, 6-Aminonicotinic acid methyl ester, 6-Aminopyridine-3-carboxylic acid methyl ester, Methyl 2-aminopyridine-5-carboxylate, Methyl 6-aminonicotinate, Methyl 6-aminopyridine-3-carboxylate |
| InChI Formula | InChI=1S/C7H8N2O2/c1-11-7(10)5-2-3-6(8)9-4-5/h2-4H,1H3,(H2,8,9) |
| SMILES | COC(=O)c1ccc(N)nc1 |
| IUPAC Name | methyl 6-aminopyridine-3-carboxylate |
| Molecular Weight (g/mol) | 152.15 |
| Formula Weight | 152.0586 |