Learn More
Methyl 5-methyl-1H-pyrazole-3-carboxylate, 97%, BLD Pharm™
Spezifikation
Spezifikation
| CAS | 25016-17-5 |
| Molecular Formula | C6H8N2O2 |
| MDL Number | MFCD03778987 |
| Quantity | 1 g |
| SMILES | COC(=O)C1=NNC(C)=C1 |
| Molecular Weight (g/mol) | 140.14 |
| Percent Purity | 97% |
| Chemical Name or Material | Methyl 5-methyl-1H-pyrazole-3-carboxylate |