missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-hexynoate, 97%

Product Code. p-7035651
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15430118

Brand: Thermo Scientific Alfa Aesar H53370.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 77758-51-1 | C7H10O2 | 126.155 g/mol

Methyl 5-hexynoate is used in the synthesis of methyl (Z)-8-chlorooct-7-ene-5-ynoate with (Z)-1,2-dichloro-ethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 5-hexynoate is used in the synthesis of methyl (Z)-8-chlorooct-7-ene-5-ynoate with (Z)-1,2-dichloro-ethane.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77758-51-1
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD00671366
InChI Key LZULAZTXJLWELL-UHFFFAOYSA-N
Synonym methyl 5-hexynoate, 5-hexynoic acid, methyl ester, 5-hexynoic acid methyl ester, hex-5-ynoic acid methyl ester, methyl 5-hexynoate #, hex-5-ynoic acid, methyl ester
PubChem CID 534077
IUPAC Name methyl hex-5-ynoate
SMILES COC(=O)CCCC#C

Specifications

Boiling Point 60°C to 62°C (10 mmHg)
Linear Formula HC≡C(CH2)3CO2CH3
Refractive Index 1.434
Quantity 5 g
Formula Weight 126.16
Percent Purity 97%
Chemical Name or Material Methyl 5-hexynoate
compliance-icons
Product Identifier
  • Methyl 5-hexynoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Methyl 5-hexynoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.