Learn More
Methyl 5-Bromovalerate, TRC
Product Code. 30482627
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482627 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30482627 Supplier Toronto Research Chemicals Supplier No. TRCM2960335G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl 5-Bromovalerate
CAS 5454-83-1
Molecular Formula C6H11BrO2
Quantity 5 g
Synonym 5-Bromopentanoic Acid Methyl Ester, 5-Bromovaleric Acid Methyl Ester, Methyl 5-Bromopentanoate, Methyl delta-Bromovalerate, Methyl ω-Bromopentanoate, NSC 23228
InChI Formula InChI=1S/C6H11BrO2/c1-9-6(8)4-2-3-5-7/h2-5H2,1H3
SMILES COC(=O)CCCCBr
IUPAC Name methyl 5-bromopentanoate
Molecular Weight (g/mol) 195.05
Formula Weight 193.9942