missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromo-2-methoxynicotinate, 97%, Thermo Scientific™

Product Code. 11444470
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444470 1 g 1g
11434470 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444470 Supplier Thermo Scientific Alfa Aesar Supplier No. H50066.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122433-41-4
Molecular Formula C8H8BrNO3
Molecular Weight (g/mol) 246.06
MDL Number MFCD08064227
InChI Key OLQOVEOEHQDKSC-UHFFFAOYSA-N
PubChem CID 14371729
IUPAC Name methyl 5-bromo-2-methoxypyridine-3-carboxylate
SMILES COC(=O)C1=CC(Br)=CN=C1OC

Specifications

Quantity 1 g
Synonym methyl 5-bromo-2-methoxynicotinate, 5-bromo-2-methoxy-nicotinic acid methyl ester, methyl 5-bromo-2-methoxy-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 5-bromo-2-methoxy-, methyl ester, 3-pyridinecarboxylicacid, 5-bromo-2-methoxy-, methyl ester, pubchem22054, acmc-209akv, methyl5-bromo-2-methoxynicotinate, methyl 5-bromo-2-methoxypyridine-3-carboxylate
Formula Weight 246.06
Percent Purity 97%
Chemical Name or Material Methyl 5-bromo-2-methoxynicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.