Learn More
Methyl 5-Aminopentanoate Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 5-Aminopentanoate Hydrochloride |
| CAS | 29840-56-0 |
| Molecular Formula | C6H13NO2 . (HCl) |
| Quantity | 1 g |
| Synonym | Methyl 5-Aminovalerate Hydrochloride |
| InChI Formula | InChI=1S/C6H13NO2.ClH/c1-9-6(8)4-2-3-5-7;/h2-5,7H2,1H3;1H |
| SMILES | Cl.COC(=O)CCCCN |
| IUPAC Name | methyl 5-aminopentanoate;hydrochloride |
| Molecular Weight (g/mol) | 167.63 |
| Formula Weight | 167.0713 |