Learn More
Methyl 4-Ethyloxybenzoate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 4-Ethyloxybenzoate |
| CAS | 23676-08-6 |
| Molecular Formula | C10H12O3 |
| Quantity | 500 mg |
| Synonym | Methyl p-Ethoxybenzoate, NSC 8378, p-Ethoxy-,methyl-benzoic Acid Ester |
| InChI Formula | InChI=1S/C10H12O3/c1-3-13-9-6-4-8(5-7-9)10(11)12-2/h4-7H,3H2,1-2H3 |
| SMILES | CCOc1ccc(cc1)C(=O)OC |
| IUPAC Name | methyl 4-ethoxybenzoate |
| Molecular Weight (g/mol) | 180.2 |
| Formula Weight | 180.0786 |