missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-cyano-5-(methylthio)thiophene-2-carboxylate, 97%, Thermo Scientific Chemicals

Product Code. 15447575
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15447575 250 mg 250mg
11431260 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15447575 Supplier Thermo Scientific Alfa Aesar Supplier No. H26353.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 175202-48-9 | Molecular Formula: C8H7NO2S2 | Molecular Weight: 213.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175202-48-9
Molecular Formula C8H7NO2S2
Molecular Weight (g/mol) 213.27
MDL Number MFCD00173757
InChI Key MJOQLNYXSSXZDJ-UHFFFAOYSA-N
Synonym methyl 4-cyano-5-methylthio thiophene-2-carboxylate, methyl 4-cyano-5-methylsulfanyl thiophene-2-carboxylate, 4-cyano-5-methylsulfanylthiophene-2-carboxylic acid methyl ester, methyl 4-cyano-5-methylthiothiophene-2-carboxylate, 2-thiophenecarboxylicacid, 4-cyano-5-methylthio-, methyl ester, acmc-20akgh, methyl 3-cyano-2-methylthio thiophene-5-carboxylate, methyl 4-cyano-5-methylsulfanyl-2-thiophenecarboxylate, 4-cyano-5-methylthio thiophene-2-carboxylic acid methyl ester
PubChem CID 18536976
IUPAC Name methyl 4-cyano-5-methylsulfanylthiophene-2-carboxylate
SMILES COC(=O)C1=CC(C#N)=C(SC)S1

Specifications

Melting Point 117°C to 119°C
Quantity 250 mg
UN Number UN3439
Formula Weight 213.27
Percent Purity 97%
Chemical Name or Material Methyl 4-cyano-5-(methylthio)thiophene-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.