Learn More
Methyl 4-(Bromomethyl)-3-methoxybenzoate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 4-(Bromomethyl)-3-methoxybenzoate |
| CAS | 70264-94-7 |
| Molecular Formula | C10 H11 Br O3 |
| Quantity | 5 g |
| Synonym | 3-Methoxy-4-(bromomethyl)benzoic Acid Methyl Ester, 4-(Bromomethyl)-3-methoxybenzoic Acid Methyl Ester, Methyl 4-(2-Bromoethyl)-3-methoxybenzoate, Methyl 4-(Bromomethyl)-3-(methyloxy)benzoate |
| InChI Formula | InChI=1S/C10H11BrO3/c1-13-9-5-7(10(12)14-2)3-4-8(9)6-11/h3-5H,6H2,1-2H3 |
| SMILES | COC(=O)c1ccc(CBr)c(OC)c1 |
| IUPAC Name | methyl 4-(bromomethyl)-3-methoxybenzoate |
| Molecular Weight (g/mol) | 259.096 |
| Formula Weight | 257.989 |