missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4,5-dibromo-3-hydroxythiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. p-3486229
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
This item is not returnable. View return policy

Product Code. 10249852

Brand: Thermo Scientific Maybridge GK01947DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 96232-71-2
Molecular Formula C6H4Br2O3S
Molecular Weight (g/mol) 315.96
MDL Number MFCD00052082
InChI Key YZPNGCXFJCPJSV-XQRVVYSFSA-N
Synonym methyl4,5-dibromo-3-hydroxythiophene-2-carboxylate, 4,5-dibromo-3-hydroxythiophene-2-carboxylic acid methyl ester, 2z-4,5-dibromo-2-hydroxy methoxy methylidene thiophen-3-one, acmc-20akgp, maybridge1_003706, methyl 4,5-dibromo-3-hydroxy-2-thiophenecarboxylate, 4,5-dibromo-2-hydroxy methoxy methylidene thiophen-3-one, methyl 4,5-dibromo-3-hydroxythiophene-2-carboxylat, 2-thiophenecarboxylicacid,4,5-dibromo-3-hydroxy-,methyl ester, 2e-4,5-dibromo-2-hydroxy methoxy methylidene thiophen-3-one
PubChem CID 2777613
IUPAC Name methyl 4,5-dibromo-3-hydroxythiophene-2-carboxylate
SMILES CO\C(O)=C1/SC(Br)=C(Br)C1=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 315.97
Percent Purity 97%
Chemical Name or Material Methyl 4, 5-dibromo-3-hydroxythiophene-2-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.