Learn More
Methyl 3-Methyl-2-butenoate, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Methyl 3-Methyl-2-butenoate |
| CAS | 924-50-5 |
| Molecular Formula | C6H10O2 |
| Quantity | 25 g |
| Synonym | Senecioic Acid Methyl Ester, 3-Methyl-2-butenoic Acid Methyl Ester, 3-Methylcrotonic Acid Methyl Ester, Methyl 3,3-Dimethacrylate, Methyl 3,3-Dimethylacrylate, Methyl 3-Methylcrotonate, Methyl Dimethylacrylate, Methyl Senecioate, Methyl beta,beta-Dimethylacrylate, Methyl beta-Methylcrotonate |
| InChI Formula | InChI=1S/C6H10O2/c1-5(2)4-6(7)8-3/h4H,1-3H3 |
| SMILES | COC(=O)C=C(C)C |
| IUPAC Name | methyl 3-methylbut-2-enoate |
| Molecular Weight (g/mol) | 114.14 |
| Formula Weight | 114.0681 |