missing translation for 'onlineSavingsMsg'
Learn More

WDFY3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. p-7235261 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18350614 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18350614 Supplier Novus Biologicals Supplier No. NBP309286100UL
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

WDFY3 Polyclonal specifically detects WDFY3 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDFY3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS buffer, 2% sucrose
Gene Alias ALFY, Autophagy-linked FYVE protein, KIAA0993MGC16461, WD repeat and FYVE domain containing 3, WD repeat and FYVE domain-containing protein 3, ZFYVE25
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of human WDFY3 (NP_848698). Peptide sequence LEVMVNLLHKYAALLKDPTQALNEQGDSRNNSSVEDQKHLALLVMETLTV
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23001
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.