Learn More
Methyl-3-keto-5Beta-cholan-24-oate, TRC
Product Code. 30483886
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483886 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30483886 Supplier Toronto Research Chemicals Supplier No. TRCM31502525MG
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl-3-keto-5beta-cholan-24-oate
CAS 1173-32-6
Molecular Formula C25H40O3
Quantity 25 mg
Synonym 3-Oxo-5beta-cholan-24-oic Acid Methyl Ester, Methyl 3-Oxo-5beta-cholan-24-oate, Methyl 3-Oxo-5beta-cholanate, 3-Oxo-5beta-cholanic Acid Methyl Ester
InChI Formula InChI=1S/C25H40O3/c1-16(5-10-23(27)28-4)20-8-9-21-19-7-6-17-15-18(26)11-13-24(17,2)22(19)12-14-25(20,21)3/h16-17,19-22H,5-15H2,1-4H3/t16-,17-,19+,20?,21+,22+,24+,25-/m1/s1
SMILES COC(=O)CC[C@@H](C)C1CC[C@H]2[C@@H]3CC[C@@H]4CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name methyl (4R)-4-[(5R,8R,9S,10S,13R,14S)-10,13-dimethyl-3-oxo-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]pentanoate
Molecular Weight (g/mol) 388.58
Formula Weight 388.2977