missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(bromomethyl)benzoate, 95%, Maybridge™

Product Code. p-4510590
Click to view available options
Quantity:
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
25g
This item is not returnable. View return policy

Product Code. 10684205

Brand: Thermo Scientific Maybridge TL00214EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1129-28-8
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00051437
InChI Key YUHSMQQNPRLEEJ-UHFFFAOYSA-N
Synonym methyl 3-bromomethyl benzoate, m-carbomethoxybenzyl bromide, 3-bromomethyl benzoic acid methyl ester, benzoic acid, 3-bromomethyl-, methyl ester, methyl-3-bromomethylbenzoate, methyl alpha-bromo-m-toluate, methyl3-bromomethyl benzoate, methyl-3-bromomethyl benzoate, 3-carbomethoxybenzyl bromide
PubChem CID 517981
IUPAC Name methyl 3-(bromomethyl)benzoate
SMILES COC(=O)C1=CC=CC(CBr)=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula BrCH2C6H4CO2CH3
Quantity 10 g
Formula Weight 229.07
Percent Purity 95%
Chemical Name or Material Methyl 3-(bromomethyl)benzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.