missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-4-cyano-5-(methylthio)thiophene-2-carboxylate, 97%, Thermo Scientific Chemicals

Product Code. p-7028824
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11366679 1 g 1g
11376679 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11366679 Supplier Thermo Scientific Alfa Aesar Supplier No. H26233.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 129332-45-2 | Molecular Formula: C8H8N2O2S2 | Molecular Weight: 228.284 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129332-45-2
Molecular Formula C8H8N2O2S2
Molecular Weight (g/mol) 228.284
MDL Number MFCD00052752
InChI Key VBXBCNWDAJLZOC-UHFFFAOYSA-N
Synonym methyl 3-amino-4-cyano-5-methylthio thiophene-2-carboxylate, methyl 3-amino-4-cyano-5-methylsulfanyl thiophene-2-carboxylate, methyl3-amino-4-cyano-5-methylthio thiophene-2-carboxylate, 2-thiophenecarboxylicacid, 3-amino-4-cyano-5-methylthio-, methyl ester, 3-amino-4-cyano-5-methylsulfanylthiophene-2-carboxylic acid methyl ester, methyl 3-azanyl-4-cyano-5-methylsulfanyl-thiophene-2-carboxylate, 3-amino-4-cyano-5-methylthio-2-thiophenecarboxylic acid methyl ester, maybridge1_009000, acmc-1c12n, methyl 3-amino-4-cyano-5-methylthiothiophene-2-carboxylate
PubChem CID 2778604
IUPAC Name methyl 3-amino-4-cyano-5-methylsulfanylthiophene-2-carboxylate
SMILES COC(=O)C1=C(C(=C(S1)SC)C#N)N

Specifications

Melting Point 205°C to 208°C
Quantity 1 g
UN Number UN3439
Formula Weight 228.28
Percent Purity 97%
Chemical Name or Material Methyl 3-amino-4-cyano-5-(methylthio)thiophene-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.