Learn More
Methyl 3,3-dimethylacrylate, 98%
Product Code. 11315585
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. 11315585
Brand: Thermo Scientific Alfa Aesar L19731.18
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 924-50-5 | C6H10O2 | 114.144 g/mol
Methyl 3,3-dimethylacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 3,3-dimethylacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Soluble in chloroform. Insoluble in water,
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. It is sensitive to light.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 924-50-5
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00043940
InChI Key FZIBCCGGICGWBP-UHFFFAOYSA-N
Synonym methyl 3-methyl-2-butenoate, methyl 3,3-dimethylacrylate, 2-butenoic acid, 3-methyl-, methyl ester, 3-methyl-but-2-enoic acid methyl ester, methyl senecioate, 3,3-dimethylacrylic acid methyl ester, methyl-3,3-dimethylacrylate, crotonic acid, 3-methyl-, methyl ester, methyl-3,3-dimethyl acrylate, methylsenecioate
PubChem CID 13546
IUPAC Name methyl 3-methylbut-2-enoate
SMILES CC(=CC(=O)OC)C
Specifications
Melting Point -41°C
Density 0.936
Boiling Point 134°C to 139°C
Flash Point 33°C (91°F)
Linear Formula (CH3)2C=CHCO2CH3
Refractive Index 1.4375
Quantity 50 g
UN Number UN3272
Beilstein 1741592
Sensitivity Light sensitive
Solubility Information Soluble in chloroform. Insoluble in water,
Formula Weight 114.14
Percent Purity 98%
Chemical Name or Material Methyl 3,3-dimethylacrylate