Learn More
Methyl 2-O-Benzoyl-4,6-di-O-benzylidene-Alpha-D-glucopyranoside, TRC
Product Code. 30483511
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483511 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30483511 Supplier Toronto Research Chemicals Supplier No. TRCM288965100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl 2-O-Benzoyl-4,6-di-O-benzylidene-alpha-D-glucopyranoside
CAS 28642-64-0
Molecular Formula C21H22O7
Quantity 100 mg
Synonym Methyl 4,6-O-(Phenylmethylene)-alpha-D-glucopyranoside 2-Benzoate
InChI Formula InChI=1S/C21H22O7/c1-24-21-18(27-19(23)13-8-4-2-5-9-13)16(22)17-15(26-21)12-25-20(28-17)14-10-6-3-7-11-14/h2-11,15-18,20-22H,12H2,1H3/t15-,16+,17-,18-,20-,21+/m1/s1
SMILES CO[C@H]1O[C@@H]2CO[C@H](O[C@H]2[C@H](O)[C@H]1OC(=O)c3ccccc3)c4ccccc4
IUPAC Name [(2R,4aR,6S,7R,8S,8aS)-8-hydroxy-6-methoxy-2-phenyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-7-yl] benzoate
Molecular Weight (g/mol) 386.4
Formula Weight 386.1366