Learn More
Methyl 2-bromobenzoate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 2-bromobenzoate |
| CAS | 610-94-6 |
| Molecular Formula | C8 H7 Br O2 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C8H7BrO2/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5H,1H3 |
| SMILES | COC(=O)c1ccccc1Br |
| IUPAC Name | methyl 2-bromobenzoate |
| Molecular Weight (g/mol) | 215.044 |
| Formula Weight | 213.963 |