Learn More
Methyl 2-Bromo-6-methoxybenzoate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 2-Bromo-6-methoxybenzoate |
| CAS | 31786-46-6 |
| Molecular Formula | C9H9BrO3 |
| Quantity | 1 g |
| Synonym | 2-Bromo-6-methoxybenzoic Acid Methyl Ester, 6-Bromo-o-anisic Acid Methyl Ester |
| InChI Formula | InChI=1S/C9H9BrO3/c1-12-7-5-3-4-6(10)8(7)9(11)13-2/h3-5H,1-2H3 |
| SMILES | O=C(OC)C(C(OC)=CC=C1)=C1Br |
| IUPAC Name | methyl 2-bromo-6-methoxybenzoate |
| Molecular Weight (g/mol) | 245.07 |
| Formula Weight | 243.97 |