Learn More
Methyl 2-Bromo-6-methoxybenzoate, TRC
Product Code. 30481689
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481689 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30481689 Supplier Toronto Research Chemicals Supplier No. TRCM29509025G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl 2-Bromo-6-methoxybenzoate
CAS 31786-46-6
Molecular Formula C9H9BrO3
Quantity 25 g
Synonym 2-Bromo-6-methoxybenzoic Acid Methyl Ester, 6-Bromo-o-anisic Acid Methyl Ester
InChI Formula InChI=1S/C9H9BrO3/c1-12-7-5-3-4-6(10)8(7)9(11)13-2/h3-5H,1-2H3
SMILES O=C(OC)C(C(OC)=CC=C1)=C1Br
IUPAC Name methyl 2-bromo-6-methoxybenzoate
Molecular Weight (g/mol) 245.07
Formula Weight 243.97