missing translation for 'onlineSavingsMsg'
Learn More

Methyl 10-undecynoate, 96%, Thermo Scientific™

Product Code. 15470278
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470278 5 g 5g
15480278 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15470278 Supplier Thermo Scientific Alfa Aesar Supplier No. H53453.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2777-66-4
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00190208
InChI Key TXFNJLOAXFOJGK-UHFFFAOYSA-N
Synonym methyl 10-undecynoate, 10-undecynoic acid, methyl ester, 10-undecynoic acid,methyl ester
PubChem CID 554128
IUPAC Name methyl undec-10-ynoate
SMILES COC(=O)CCCCCCCCC#C

Specifications

Density 0.908
Boiling Point 124°C (10 mmHg)
Linear Formula HC≡C(CH2)8CO2CH3
Refractive Index 1.4464
Quantity 5 g
Formula Weight 196.29
Percent Purity 96%
Chemical Name or Material Methyl 10-undecynoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.